
1,4-Dibromobenzene ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:106-37-6
:
Formula:C₆H₄Br₂
MDL Number:MFCD00000089
Merck Index:14,03019
Boiling Pt:219°C
Melting Pt:84…87 °C
Density:2,28 g/cm³ (20 °C)
Flash Pt:≥ 100 °C
ADR:9,III
:235.91 g/mol
EINECS:203-390-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Light yellow powder to lump
- Purity (GC):min. 99.0 %
- Melting point:88.0 to 90.0 deg-C
Specification Test Results
Frequently Bought Together