
1,2-Dibromobenzene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:583-53-9
:
Formula:C₆H₄Br₂
MDL Number:MFCD00000057
:235.91 g/mol
Boiling Pt:224°C
Density:1,956 g/cm³ (20 °C)
Flash Pt:91°C
Storage Temperature:Refrigerator
EINECS:209-507-3
ADR:9,III
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together