
Methyl-2-amino-4,5-dimethoxybenzoate ≥98.0% (by GC, titration analysis)
TCIAD2749-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:128.0 to 131.0 deg-C
:
:211.22 g/mol
:247-967-7
:GHS07
:26759-46-6
:C₁₀H₁₃NO₄
:MFCD00014904