Order Entry
Italy
ContactUsLinkComponent
Methyl-5-chloro-2-methoxybenzoate ≥97.0% (by GC)
Methyl-5-chloro-2-methoxybenzoate ≥97.0% (by GC)
Catalog # TCIAC1457-25G
Supplier:  TCI
CAS Number:  
undefined
Methyl-5-chloro-2-methoxybenzoate ≥97.0% (by GC)
Catalog # TCIAC1457-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :200.62 g/mol
Boiling Pt:240°C
EINECS:251-743-4
CAS Number:33924-48-0
Formula:C₉H₉ClO₃
MDL Number:MFCD00017511