0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
2,5-Diacetoxytoluene
2,5-Diacetoxytoluene ≥97.0% (by GC)
2,5-Diacetoxytoluene ≥97.0% (by GC)
TCIAD3318-25G
:
TCI
:
2,5-Diacetoxytoluene ≥97.0% (by GC)
TCIAD3318-25G
:
TCI
:
Pack type:
Glass bottle
Pk:
25 g
Seq:
3308
Appearance:
White to Light yellow powder to crystal
Purity (GC):
min. 97.0 %
:
:
208.21
:
49°C
:
717-27-1
:
C₁₁H₁₂O₄
:
MFCD00061098