trans-1,4-Diphenyl-2-butene-1,4-dione ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:959-28-4
:
Formula:C₁₆H₁₂O₂
MDL Number:MFCD00003083
:236.27 g/mol
EINECS:213-498-1
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:109.0 to 112.0 deg-C
Specification Test Results
Frequently Bought Together