Order Entry
Italy
ContactUsLinkComponent
(1S,2S)-(-)-1,2-Diphenylethylenediamine ≥98.0% (by GC, titration analysis)
Irritant
(1S,2S)-(-)-1,2-Diphenylethylenediamine ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
(1S,2S)-(-)-1,2-Diphenylethylenediamine ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    81.0 to 85.0 deg-C
  • Specific rotation [a]20/D:
    -98.0 to -108.0 deg (C=1, EtOH)

Specification Test Results

Frequently Bought Together