1,8-Dichlorohexadecafluorooctane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:647-25-6
:
Formula:C₈Cl₂F₁₆
MDL Number:MFCD00155747
:470.97 g/mol
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Specific gravity (20/20):1.7980 to 1.8080
- Refractive index n20/D:1.3120 to 1.3160
Specification Test Results
Frequently Bought Together