1,2-Dibenzoylethane ≥98.0% (by GC)
Catalog # TCIAD3179-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (GC):min. 98.0 %
- Melting point:145.0 to 149.0 deg-C
- Solubility in hot Acetone:almost transparency
Specification Test Results
About this item
:
:238.29 g/mol
Boiling Pt:260°C
CAS Number:495-71-6
Formula:C₁₆H₁₄O₂
MDL Number:MFCD00037818