Order Entry
Italy
ContactUsLinkComponent
2,3-Dimethyl-1-pentene ≥98.0% (by GC)
2,3-Dimethyl-1-pentene ≥98.0% (by GC)
Catalog # TCIAD1248-1ML
Supplier:  TCI
CAS Number:  
undefined
2,3-Dimethyl-1-pentene ≥98.0% (by GC)
Catalog # TCIAD1248-1ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
Boiling Pt:84°C
Melting Pt:–134.3 °C
Density:0,7051 g/cm³ (20 °C)
Flash Pt:-10°C
Flash Pt:–10°C
Storage Temperature:Ambient
EINECS:222-285-2
UN:3295
ADR:3,II
 :98.19 g/mol
 :GHS02
 :GHS08
CAS Number:3404-72-6
Formula:C₇H₁₄
MDL Number:MFCD00027075