trans-2-Pentene ≥99.0% (by GC)
TCIAP0635-10ML
: TCI
:
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
- Environmentally Preferable:
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
: (E)-2-Pentene
:
:14,07123
:36°C
:–140.2 °C
:0,6431 g/cm³ (25 °C)
:-45°C
:–45°C
:Ambient
:211-461-4
:3295
:3,II
:70.13 g/mol
:GHS02
:GHS07
:GHS08
:646-04-8
:C₅H₁₀
:MFCD00009384