

cis-2-Hexene ≥95.0% (by GC)
Catalog # TCIAH0404-5ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
About this item
:
:84.16 g/mol
Boiling Pt:69°C
Melting Pt:–141.1 °C
Density:0,6863 g/cm³ (20 °C)
Flash Pt:-20°C
Flash Pt:–20°C
Storage Temperature:Ambient
EINECS:231-697-1
UN:3295
ADR:3,II
:GHS02
:GHS08
CAS Number:7688-21-3
Formula:C₆H₁₂
MDL Number:MFCD00066520