Order Entry
Italy
Orders LinkContactUsLinkComponent
 

1-Hexene ≥97.0% (by GC)

Flammable
Irritant
Health
Supplier:   TCI
1-Hexene ≥97.0% (by GC)
Supplier:  TCI
Certificates
1-Hexene ≥97.0% (by GC)

2 Options Available Below

Product Details & Documents
  • CAS Number:592-41-6
  • Formula:C₆H₁₂
  • MDL Number:MFCD00009505
  • Molecular Weight:84.16 g/mol
  • Boiling Pt:64°C
  • Melting Pt:–140°C
  • Melting Pt:-140°C
  • Density:0,675 g/cm³ (20 °C)
  • Flash Pt:–31°C
  • Flash Pt:-31°C
  • Storage Temperature:Ambient
  • EINECS:209-753-1
  • UN:2370
  • ADR:3,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 17,00 €
    17,00 €
    1 * 25 mL
       17,00 €

  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 28,00 €
    28,00 €
    1 * 500 mL
       28,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.