Order Entry
Italy
ContactUsLinkComponent
Acrolein dimethyl acetal ≥97.0% (by GC)
Flammable
Irritant
Acrolein dimethyl acetal ≥97.0% (by GC)
Catalog # TCIAA2455-250G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Acrolein dimethyl acetal ≥97.0% (by GC)
Catalog # TCIAA2455-250G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :102.13 g/mol
Boiling Pt:90°C
Density:0,862 g/cm³ (20 °C)
Flash Pt:-3°C
Flash Pt:–3°C
EINECS:227-936-4
UN:1993
ADR:3,II
 :GHS02
 :GHS07
CAS Number:6044-68-4
Formula:C₅H₁₀O₂
MDL Number:MFCD00008632