Order Entry
Italy
ContactUsLinkComponent
2,3-Dichloropropene ≥97.0% (by GC)
Flammable
Corrosive
Irritant
Health
2,3-Dichloropropene ≥97.0% (by GC)
Catalog # TCIAD0406-25G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
2,3-Dichloropropene ≥97.0% (by GC)
Catalog # TCIAD0406-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :110.97 g/mol
Boiling Pt:94°C
Melting Pt:–83.1 °C
Density:1,209 g/cm³ (20 °C)
Flash Pt:10°C
Storage Temperature:Refrigerator
EINECS:201-153-8
UN:2047
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:78-88-6
Formula:C₃H₄Cl₂
MDL Number:MFCD00000943