

5-Bromo-1-pentene ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1119-51-3
:
Formula:C₅H₉Br
MDL Number:MFCD00000264
:149.03 g/mol
Boiling Pt:126°C
Density:1,2547 g/cm³ (15 °C)
Flash Pt:30°C
Storage Temperature:Refrigerator
EINECS:214-281-4
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together