Allyl bromide ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:106-95-6
:
Formula:C₃H₅Br
MDL Number:MFCD00000244
:120.98 g/mol
Merck Index:14,00288
Boiling Pt:71°C
Melting Pt:–119 °C
Density:1,4 g/cm³ (20 °C)
Flash Pt:12°C
EINECS:203-446-6
UN:1099
ADR:3,I
:GHS02
:GHS05
:GHS06
:GHS09
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together