Order Entry
Italy
ContactUsLinkComponent
2-Bromopropene ≥97.0% (by GC)
2-Bromopropene ≥97.0% (by GC)
Catalog # TCIAB1269-10G
Supplier:  TCI
CAS Number:  
undefined
2-Bromopropene ≥97.0% (by GC)
Catalog # TCIAB1269-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

Synonyms: 2-Propenyl bromide, α-Methylvinyl bromide, Isopropenyl bromide
 :
 :120.98 g/mol
Boiling Pt:48°C
Melting Pt:–125 °C
Density:1,39 g/cm³ (9 °C)
Flash Pt:4°C
Storage Temperature:Refrigerator
EINECS:209-185-4
UN:1993
ADR:3,II
 :GHS02
 :GHS07
CAS Number:557-93-7
Formula:C₃H₅Br
MDL Number:MFCD00000140