Order Entry
Italy
ContactUsLinkComponent
2,3-Dibromopropene ≥95.0% (by GC)
Irritant
2,3-Dibromopropene ≥95.0% (by GC)
Catalog # TCIAD0205-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2,3-Dibromopropene ≥95.0% (by GC)
Catalog # TCIAD0205-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :199.87 g/mol
Merck Index:14,03025
Boiling Pt:141°C
Density:2,05 g/cm³ (20 °C)
Flash Pt:81°C
EINECS:208-155-8
 :GHS07
Storage Temperature:Freezer
CAS Number:513-31-5
Formula:C₃H₄Br₂
MDL Number:MFCD00000211