2,6,10,14-Tetramethylpentadecane ≥95.0% (by GC)
TCIAT0149-5ML
: TCI
:
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
:
:268.53 g/mol
:14,07752
:299°C
:-99°C
:–99°C
:0,7845 g/cm³ (20 °C)
:Ambient
:217-650-8
:GHS07
:1921-70-6
:C₁₉H₄₀
:MFCD00008952