2-Chloro-2-methylbutane ≥97.0% (by GC)
: TCI
:594-36-5
:
:C₅H₁₁Cl
:MFCD00000818
:86°C
:–73 °C
:0,8591 g/cm³ (20 °C)
:–9°C
:Ambient
:209-836-2
:3,II
:106.6 g/mol
:1107
:GHS02
:GHS07
- Environmentally Preferable:
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
- Specific gravity (20/20):0.8610 to 0.8680
- Refractive index n20/D:1.4040 to 1.4060
Frequently Bought Together