

1-Chloro-2,2-dimethylpropane ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:753-89-9
:
Formula:C₅H₁₁Cl
MDL Number:MFCD00000941
:106.6 g/mol
Boiling Pt:84 °C (1013 hPa)
Melting Pt:–20 °C
Density:0,86 g/cm³ (20 °C)
Flash Pt:–8°C
EINECS:212-040-8
UN:1993
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together