
Pentaerythrityl tetrachloride ≥98.0% (by GC)
: TCI
:3228-99-7
:
:C₅H₈Cl₄
:MFCD00000942
:110°C
:95 °C
:1,675 g/cm³ (20 °C)
:209.93 g/mol
:221-763-8
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:95.0 to 98.0 deg-C
- Solubility in Methanol:very faint turbidity
Frequently Bought Together