Order Entry
Italy
ContactUsLinkComponent
4-Methyl-2-pentanone ≥99.5% (by GC)
Flammable
Irritant
4-Methyl-2-pentanone ≥99.5% (by GC)
Supplier:  TCI
Certificates
undefined
Flammable
Irritant
4-Methyl-2-pentanone ≥99.5% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.5 %

Specification Test Results

Frequently Bought Together