
n-Tetradecane ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:629-59-4
:
Formula:H₃C(CH₂)₁₂CH₃
MDL Number:MFCD00008986
:198.39 g/mol
Boiling Pt:253°C
Melting Pt:6°C
Density:0,7628 g/cm³ (20 °C)
Flash Pt:100°C (closed cup)
EINECS:211-096-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together