4,4,4-Trifluoro-1-phenyl-1,3-butanedione ≥98.0% (by GC)
Catalog # TCIAT0437-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to lump
- Purity (GC):min. 98.0 %
- Melting point:37.0 to 41.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:216.16
Boiling Pt:224°C
EINECS:206-307-8
CAS Number:326-06-7
Formula:C₁₀H₇F₃O₂
MDL Number:MFCD00000425