Order Entry
Italy
ContactUsLinkComponent
1H,1H,11H-Eicosafluoro-1-undecanol ≥90.0% (by GC)
1H,1H,11H-Eicosafluoro-1-undecanol ≥90.0% (by GC)
Catalog # TCIAE0239-10G
Supplier:  TCI
CAS Number:  
undefined
1H,1H,11H-Eicosafluoro-1-undecanol ≥90.0% (by GC)
Catalog # TCIAE0239-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 90.0 %
  • Melting point:
    94.0 to 98.0 deg-C

Specification Test Results

About this item

 :
 :532.12 g/mol
EINECS:206-209-5
CAS Number:307-70-0
Formula:C₁₁H₄F₂₀O
MDL Number:MFCD00039628