Order Entry
Italy
ContactUsLinkComponent
1,6-Hexanedithiol ≥97.0% (by GC, titration analysis)
Irritant
1,6-Hexanedithiol ≥97.0% (by GC, titration analysis)
Catalog # TCIAH0334-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
1,6-Hexanedithiol ≥97.0% (by GC, titration analysis)
Catalog # TCIAH0334-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %
  • Purity (Iodometric Titration):
    min. 97.0 %

Specification Test Results

About this item

 :
Boiling Pt:237°C
Melting Pt:–21 °C
Density:1,0102 g/cm³ (0 °C)
Flash Pt:90°C
EINECS:214-735-1
ADR:6.1,III
 :150.31 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1191-43-1
Formula:C₆H₁₄S₂
MDL Number:MFCD00004910