
1,4-Butanedithiol ≥95.0% (by GC, titration analysis)
: TCI
:1191-08-8
:
:C₄H₁₀S₂
:MFCD00004906
:122.26 g/mol
:195°C
:–53 °C
:1,0621 g/cm³ (0 °C)
:70°C
:Ambient
:214-728-3
:9,
:GHS07
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
- Purity (Iodometric Titration):min. 95.0 %
Frequently Bought Together