Order Entry
Italy
ContactUsLinkComponent
1-Hexanol ≥98.0% (by GC)
Flammable
Irritant
1-Hexanol ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Flammable
Irritant
1-Hexanol ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together