1-Heneicosanol ≥98.0% (by GC)
TCIAH0011-10G
: TCI
:
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
- Appearance:White powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:67.0 to 70.0 deg-C
- Solubility in hot Methanol:almost transparency
:
:312.58 g/mol
:170 °C (1013 hPa)
:68…69 °C
:Refrigerator
:239-673-2
:15594-90-8
:H₃C(CH₂)₁₉CH₂OH
:MFCD00062834