1-Docosanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:661-19-8
:
Formula:H₃C(CH₂)₂₀CH₂OH
MDL Number:MFCD00002939
:326.61 g/mol
Merck Index:13,03433
Boiling Pt:376 °C (1013 hPa)
Melting Pt:68…71 °C
Density:~ 0,81 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-546-6
Specification Test Results
- Appearance:White pellet
- Purity (GC):min. 98.0 %
- Melting point:68.0 to 73.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together