


1-Decanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:112-30-1
:
Formula:H₃C(CH₂)₈CH₂OH
MDL Number:MFCD00004747
:158.28 g/mol
Merck Index:14,02855
Boiling Pt:233°C
Melting Pt:7 °C
Density:0,82634 g/cm³ (20 °C)
Flash Pt:117°C
Storage Temperature:Ambient
EINECS:203-956-9
UN:3082
ADR:9,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together