



Triethylamine ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:121-44-8
:
Formula:(C₂H₅)₃N
MDL Number:MFCD00009051
:101.19 g/mol
Merck Index:14,09666
Boiling Pt:89°C
Melting Pt:–115°C
Density:0,729 g/cm³ (20 °C)
Flash Pt:–8°C
EINECS:204-469-4
UN:1296
ADR:3,II
:GHS02
:GHS05
:GHS06
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
- Purity (Nonaqueous Titration):min. 99.0 %
Specification Test Results
Frequently Bought Together