Order Entry
Italy
ContactUsLinkComponent
1,2-Dichloro-1-ethoxyethane ≥93.0% (by GC)
Flammable
Irritant
Health
1,2-Dichloro-1-ethoxyethane ≥93.0% (by GC)
Catalog # TCIAD0355-25G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Health
1,2-Dichloro-1-ethoxyethane ≥93.0% (by GC)
Catalog # TCIAD0355-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 93.0 %

Specification Test Results

About this item

 :
 :143.01 g/mol
Boiling Pt:145°C
Melting Pt:–72 °C
Density:1,167 g/cm³ (25 °C)
Flash Pt:35°C
EINECS:210-794-2
UN:3271
ADR:3,III
 :GHS02
 :GHS07
 :GHS08
CAS Number:623-46-1
Formula:H₃CCH₂OCClHCClH₂
MDL Number:MFCD00041730