Order Entry
Italy
ContactUsLinkComponent
Chloroform ≥99.0% (by GC) stabilised for HPLC
Toxic
Health
Environment
Chloroform ≥99.0% (by GC) stabilised for HPLC
Catalog # TCIAC0819-500ML
Supplier:  TCI
CAS Number:  
undefined
Toxic
Health
Environment
Chloroform ≥99.0% (by GC) stabilised for HPLC
Catalog # TCIAC0819-500ML
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.0 %
  • Density (20deg.C):
    1.4740 to 1.4910 g/mL
  • Absorbance (250nm):
    max. 0.500
  • Absorbance (260nm):
    max. 0.150

Specification Test Results

About this item

 :
 :119.38 g/mol
Merck Index:14,02141
Boiling Pt:61,7 °C (1013 hPa)
Melting Pt:-64°C
Melting Pt:–64°C
Density:1,4832 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
 :Stabilised with ethanol
REACH:05-2114614906-45
 :GHS06
 :GHS08
 :GHS09
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826