Order Entry
Italy
ContactUsLinkComponent
1,5-Dichloropentane ≥95.0% (by GC)
Flammable
Irritant
1,5-Dichloropentane ≥95.0% (by GC)
Catalog # TCIAD0390-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
1,5-Dichloropentane ≥95.0% (by GC)
Catalog # TCIAD0390-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :141.04 g/mol
Boiling Pt:182°C
Melting Pt:–72.8 °C
Density:1,1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-053-6
UN:1152
ADR:3,III
 :GHS02
 :GHS07
CAS Number:628-76-2
Formula:ClH₂C(CH₂)₃CH₂Cl
MDL Number:MFCD00001017