Order Entry
Italy
ContactUsLinkComponent
1,4-Dichlorobutane ≥98.0% (by GC)

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

Frequently Bought Together