


Amyl acetate ≥99.0% (by GC)
: TCI
:628-63-7
:
:C₇H₁₄O₂
:MFCD00009500
:130.19 g/mol
:148°C
:–71 °C
:0,876 g/cm³ (20 °C)
:45°C
:Ambient
:211-047-3
:1104
:3,III
:GHS02
:GHS07
:GHS08
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
Frequently Bought Together