

2-Pentanone ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:107-87-9
:
Formula:C₅H₁₀O
MDL Number:MFCD00009400
:86.13 g/mol
Merck Index:14,06114
Boiling Pt:101°C
Melting Pt:–78 °C
Density:0,809 g/cm³ (20 °C)
Flash Pt:7°C
Storage Temperature:Ambient
EINECS:203-528-1
UN:1249
ADR:3,II
:GHS02
:GHS06
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together