
1-Ethyl-3-methylpyridinium ethylsulphate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:872672-50-9
:
Formula:C₁₀H₁₇NO₄S
:247.32 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Yellow to Orange clear liquid
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together