

2-Mercaptobenzimidazole ≥98.0% (by HPLC, titration analysis)
: TCI
:583-39-1
:
:C₇H₆N₂S
:MFCD00466107
:150.2 g/mol
:14,01082
:376 °C (1013 hPa)
:154 °C
:1,42 g/cm³ (20 °C)
:209-502-6
:2811
:6.1,III
:GHS06
:GHS08
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot Methanol:almost transparency
Frequently Bought Together