
1,4-Dithioerythritol ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6892-68-8
:92-68-8
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00063750
:154.25 g/mol
Boiling Pt:286 °C (1013 hPa)
Melting Pt:82…83 °C
Storage Temperature:Refrigerator
EINECS:229-998-8
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Iodometric Titration):min. 98.0 %
- Melting point:83.0 to 86.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together