Order Entry
Italy
ContactUsLinkComponent
(±)-3-Methoxy-1,2-propanediol ≥98.0% (by GC)
(±)-3-Methoxy-1,2-propanediol ≥98.0% (by GC)
Catalog # TCIAM0125-25ML
Supplier:  TCI
CAS Number:  
undefined
(±)-3-Methoxy-1,2-propanediol ≥98.0% (by GC)
Catalog # TCIAM0125-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :106.12 g/mol
Boiling Pt:220°C
Density:1,12 g/cm³ (20 °C)
EINECS:210-791-6
CAS Number:623-39-2
Formula:C₄H₁₀O₃
MDL Number:MFCD00004719