2-Hydroxy-5-nitro-3-picoline ≥98.0% (by GC)
: TCI
: 3-Methyl-5-nitropyridin-2-ol, 3-Methyl-5-nitropyridin-2(1H)-one, 3-Methyl-5-nitro-2-pyridone, 2-Hydroxy-3-methyl-5-nitropyridine
:21901-34-8
:
:C₆H₆N₂O₃
:MFCD03095073
:154.13 g/mol
:229°C
:244-647-9
:GHS07
- Environmentally Preferable:
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (GC):min. 98.0 %
Frequently Bought Together