


Triethylammonium dihydrogentrifluoride ≥95.0% (by titrimetric analysis)
Catalog # TCIAT2022-10G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:Colorless to Red to Green clear liquid
- Purity (Nonaqueous Titration):min. 95.0 %
Specification Test Results
About this item
:
:161.21 g/mol
Density:1,094 g/cm³ (20 °C)
EINECS:277-550-5
UN:2927
ADR:6.1,I
:GHS05
:GHS06
:GHS08
CAS Number:73602-61-6
Formula:C₆H₁₈F₃N
MDL Number:MFCD00043294