Dexamethasone ≥99.0% (by HPLC)
Catalog # TCIAD1961-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 99.0 area%
- Specific rotation [a]20/D:+72.0 to +80.0 deg (C=1, Dioxane)
Specification Test Results
About this item
:
:392.47 g/mol
Merck Index:14,02943
Melting Pt:264°C
EINECS:200-003-9
:GHS08
CAS Number:50-02-2
Formula:C₂₂H₂₉FO₅
MDL Number:MFCD00064136