
2-(2-Ethoxyethoxy)ethyl acetate ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:112-15-2
:
Formula:C₈H₁₆O₄
MDL Number:MFCD00041928
:176.21 g/mol
Boiling Pt:218°C
Melting Pt:–25 °C
Density:1,0114 g/cm³ (20 °C)
Flash Pt:107°C
EINECS:203-940-1
:GHS07
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together