Order Entry
Italy
ContactUsLinkComponent
 
 
2,4-Dihydroxybenzamide ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
 
2,4-Dihydroxybenzamide ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light red to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %

 

Frequently Bought Together