Order Entry
Italy
ContactUsLinkComponent
 
 
3,4-Dimethoxy-3-cyclobutene-1,2-dione ≥98.0% (by GC)
 :  TCI
 
undefined
 
3,4-Dimethoxy-3-cyclobutene-1,2-dione ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    55.0 to 58.0 deg-C

 

Frequently Bought Together